Learn More
Invitrogen CELLSENSOR(R) IRF1-BLA BA/F3 1ml
Brand:  Invitrogen K1205
CELLSENSOR(R) IRF1-BLA BA/F3 1ml
Product Code. 10023992
  • 122300.00 SEK / 1mL
For Research Use Only. All usage must comply with product instructions.
Product Title
Invitrogen CELLSENSOR(R) IRF1-BLA BA/F3 1ml >