Learn More
Invitrogen GeneBlazer(R) EDNRB-NFAT-BLA HEK 293T CELL 1ml
Brand:  Invitrogen K1224
GeneBlazer(R) EDNRB-NFAT-BLA HEK 293T CELL 1ml
Product Code. 10013802
  • 168500.00 SEK / 1mL
For Research Use Only. All usage must comply with product instructions.
Product Title
Invitrogen GeneBlazer(R) EDNRB-NFAT-BLA HEK 293T CELL 1ml >