Learn More
TdB Labs AB 10 kDa Q-dextran for Ion-exchange Chromatography and Biomolecular Studies
Q-dextran is a positively charged dextran modified with quaternary ammonium groups. It is used in ion exchange chromatography, biochemical assays, and for studying electrostatic interactions with negatively charged biomolecules.
Encompass_Preferred
1488.48 SEK - 13532.14 SEK
Specifications
Product Type Modified Dextran
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30216327
Encompass_Preferred
View Documents
TdB Labs AB
QD10_1g
1 g
1488.48 SEK
1g
View alternative products
Save Up to 
»
30216265
Encompass_Preferred
View Documents
TdB Labs AB
QD10_10g
10 g
13532.14 SEK
10g
View alternative products
Save Up to 
»
Description
Description
  • Storage and Stability:
    • Store dry at room temperature, protected from moisture and light.
    • Stable for extended periods under recommended storage conditions without loss of charge properties.
  • Specifications
    Specifications
    Modified Dextran
    Videos
    SDS
    Documents
    Documents
    Product Certifications
    Product Content Correction
    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
    Product Title
    TdB Labs AB 10 kDa Q-dextran for Ion-exchange Chromatography and Biomolecular Studies