Learn More
VHG Labs 100ML ICP-MS CRQL Solution 1: Co. Mn at5: Ag. As.Be. Cd. Ni. Pb. Tl. V. Zn at10: Cr. Cu. Sb at 20
Brand: VHG Labs VHG-CRQL1MS-100
100ML ICP-MS CRQL Solution 1: Co. Mn at5: Ag. As.Be. Cd. Ni. Pb. Tl. V. Zn at10: Cr. Cu. Sb at 20
This item is not returnable.
View return policy
Product Content Correction
Product Title
VHG Labs 100ML ICP-MS CRQL Solution 1: Co. Mn at5: Ag. As.Be. Cd. Ni. Pb. Tl. V. Zn at10: Cr. Cu. Sb at 20 >
Spot an opportunity for improvement?Share a Content Correction