Learn More
Sartorius™ 11403Z Series Gridded Sterile Cellulose Nitrate (CN) Membrane Filters
11403Z Series Gridded Sterile Cellulose Nitrate (CN) Membrane Filters are ideal for colony counting, particle testing, and microscopy. Specifically designed for use in Microsart™ e.motion dispensers (and Sartorius nutrient pad sets), they are supplied individually sterile-sealed on a pleated band.
Brand: Sartorius™ 11403Z-47----SCM
Additional Details : Weight : 0.01000kg
Description
- High flow membranes
- Outstanding recovery rates for microorganisms
- Multi-fit: Fits into other dispensers
- Protective paper-free
- Packaged on a special pleated band
- Product data is printed on individual envelopes
- Black grid on white membrane
- Gamma irradiated, 25 kGray
Specifications
White | |
Cellulose Nitrate | |
1.2 μm | |
For colony counting, particle testing and microscopy | |
300 Pack | |
Membrane Filter | |
47 mm |
Membrane Filter | |
Cellulose Nitrate | |
Sterile | |
Black | |
115 to 145 μm | |
47 mm |
Safety and Handling
- Danger
Hazard Category
- Flammable solid Category 1
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P230-Keep wetted with .
- P233-Keep container tightly closed.
- P234-Keep only in original container.
- P243-Take precautionary measures against static discharge.
- P370+P378-In case of fire: Use to extinguish.
- P372-Explosion risk in case of fire.
- P375-Fight fire remotely due to the risk of explosion.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.