Learn More
Bochem™ 18/10 Stainless Steel Spatulas, Spoon/round/bent
Manufactured from 18/10 stainless steel
Brand: Bochem™ 3204
Additional Details : Weight : 0.02100kg
Description
- Made from 18/10 stainless steel
Specifications
1 Ea. | |
Spoon/Round/Bent | |
Used for a variety of applications | |
4mm | |
210 mm |
Autoclavable | |
Spatula | |
Stainless Steel (18/10) | |
Stainless Steel (18/10) |