Läs mer
2,3-Difluoro-4-methoxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
Produktkod. 15447386
Change view
Kvantitet:
250 mg
1 g
Förpackningsstorlek:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15447386 250 mg 250mg
15437386 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 15447386 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H62399.MD
Request Bulk or Custom Format
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1809298-92-7
Synonym 2,3-Difluoro-4-methoxyphenylboronic acid pinacol ester
Specifikationer
Kemiskt namn eller material 2,3-Difluoro-4-methoxybenzeneboronic acid pinacol ester
Kvantitet 250 mg
Formel vikt 270.08
Procent renhet 96%
Fysisk form Solid