Läs mer
2,4,6-Triamino-5-pyrimidinecarbonitrile, TRC
Produktkod. 30006581
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006581 10 mg
30006967 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30006581 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT76722010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 465531-97-9
Molekylformel C5 H6 N6
Molekylvikt (g/mol) 150.14
Synonym 2,4,6-Triamino-5-pyrimidinecarbonitrile, Dicyclanil-descyclopropyl, Descyclopropyldicyclanil
IUPAC-namn 2,4,6-triaminopyrimidine-5-carbonitrile
LEDER Nc1nc(N)c(C#N)c(N)n1
Specifikationer
Kemiskt namn eller material 2,4,6-Triamino-5-pyrimidinecarbonitrile
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C5H6N6/c6-1-2-3(7)10-5(9)11-4(2)8/h(H6,7,8,9,10,11)
Formel vikt 150.0654
Procent renhet >95%