Läs mer
2,4-Dichlorobenzamidoxime, 97%, Thermo Scientific™
Produktkod. 11410220
Kvantitet:
250 mg
1 g
Förpackningsstorlek:
1g
250mg
Produktkod. 11410220
Brand: Thermo Scientific Alfa Aesar H50674.MD
Request Bulk or Custom Format
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 22179-80-2
Molekylformel C7H6Cl2N2O
Molekylvikt (g/mol) 205.038
MDL-nummer MFCD00178762
InChI-nyckel MANQHFUAAAGDIX-UHFFFAOYSA-N
Synonym 2,4-dichlorobenzamidoxime, 2,4-dichloro-n'-hydroxybenzimidamide, 2,4-dichloro-n'-hydroxybenzene-1-carboximidamide, 2,4-dichloro-n-hydroxybenzenecarboximidamide, 2,4-dichlorophenyl hydroxyimino methylamine, z-2,4-dichloro-n'-hydroxybenzimidamide, benzenecarboximidamide,2,4-dichloro-n-hydroxy, hcmhpngpbebdfyyui``xhacp, 2,4-dichloro-n-hydroxybenzimidamide, 2,4-bis chloranyl-n'-oxidanyl-benzenecarboximidamide
PubChem CID 9581031
IUPAC-namn 2,4-dichloro-N'-hydroxybenzenecarboximidamide
LEDER C1=CC(=C(C=C1Cl)Cl)C(=NO)N
Specifikationer
Kemiskt namn eller material 2,4-Dichlorobenzamidoxime
Kvantitet 250 mg
Formel vikt 205.04
Procent renhet 97%