Learn More
2,8-Bis(trifluoromethyl)-4-hydroxyquinoline, 97%, Thermo Scientific Chemicals Product Code.: 11458627
2,8-Bis(trifluoromethyl)-4-hydroxyquinoline, 97%, Thermo Scientific Chemicals
Product Code. p-7024669
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11458627
Brand: Thermo Scientific Alfa Aesar A19750.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 35853-41-9
Molecular Formula C11H5F6NO
Molecular Weight (g/mol) 281.157
MDL Number MFCD00075091
InChI Key JIWHKBAFGFPZKM-UHFFFAOYSA-N
Synonym 2,8-bis trifluoromethyl-4-hydroxyquinoline, 2,8-bis trifluoromethyl quinolin-4-ol, 2,8-bis trifluoromethyl-4-quinolinol, 2,8-bis trifluoromethyl-1h-quinolin-4-one, 2,8-bis-trifluoromethyl-quinolin-4-ol, cc-pmlsc-dma-p104, 4-quinolinol, 2,8-bis trifluoromethyl, 2,8-bis trifluoromethyl-4-hydroxy quinoleine, 2,8-di trifluoromethyl quinolin-4-ol, pubchem5934Show MoreShow Less
PubChem CID 736140
IUPAC Name 2,8-bis(trifluoromethyl)-1H-quinolin-4-one
SMILES C1=CC2=C(C(=C1)C(F)(F)F)NC(=CC2=O)C(F)(F)F
Specifications
Melting Point 129°C to 135°C
Quantity 1 g
Formula Weight 281.16
Percent Purity 97%
Chemical Name or Material 2,8-Bis(trifluoromethyl)-4-hydroxyquinoline
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Product Code
Lot Number10133255 Certificate TypeCertificate of Analysis Date25/10/2023 Product CodeA19750.03,A19750.06
RUO – Research Use Only
Product Content Correction
Product Title
2,8-Bis(trifluoromethyl)-4-hydroxyquinoline, 97%, Thermo Scientific Chemicals >