Learn More
Julabo™ 2-Adapters for Presto™ Temperature Control System
Brand:  Julabo™ 8890083
2678.26 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15214283
  • 2725.00 SEK / Pack of 2
Only null left
Estimated Shipment: 20-08-2025
Specifications
Suitable for PRESTO A30, PRESTO A40, PRESTO A45, PRESTO A45t, PRESTO A80 Temperature Control System
2-Adapters
M30 x 1.5 Female to NPT 3/4 in. Male
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Julabo™ 2-Adapters for Presto™ Temperature Control System > Connection Type: M30 x 1.5 Female to NPT 3/4 in. Male