missing translation for 'onlineSavingsMsg'
Läs mer

2-Bromobenzaldehyde diethyl acetal, 98%, Thermo Scientific™

Produktkod. p-7046641
Click to view available options
Kvantitet:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11308296

Brand: Thermo Scientific Alfa Aesar L15513.06

Request Bulk or Custom Format

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Detta Thermo Scientific varumärkesprodukt var ursprungligen en del av Alfa Aesar produktportfölj. Viss dokumentation och etikettinformation kan referera till det äldre varumärket. Originalet Alfa Aesar produkt-/artikelkod eller SKU-referens har inte ändrats som en del av varumärkesövergången till Thermo Scientific .

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35822-58-3
Molekylformel C11H15BrO2
Molekylvikt (g/mol) 259.14
MDL-nummer MFCD01075696
InChI-nyckel APDGYQVYBWGBSD-UHFFFAOYSA-N
Synonym 2-bromobenzaldehyde diethyl acetal, 1-bromo-2-diethoxymethyl benzene, 2-diethoxymethyl bromobenzene, 2-bromobenzaldehydediethylacetal, benzene, 1-bromo-2-diethoxymethyl, 2-bromophenyl diethoxymethane, 2-bromo-alpha,alpha-diethoxytoluene, pubchem5370, acmc-1afuv, bromobenzaldehyde diethylacetal
PubChem CID 3482778
IUPAC-namn 1-bromo-2-(diethoxymethyl)benzene
LEDER CCOC(OCC)C1=CC=CC=C1Br

Specifications

Kemiskt namn eller material 2-Bromobenzaldehyde diethyl acetal
Densitet 1.285
Kokpunkt 95°C (1 mmHg)
Flampunkt 113°C (235°F)
Brytningsindex 1.515
Kvantitet 5 g
Beilstein 5935613
Formel vikt 259.14
Procent renhet 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.