missing translation for 'onlineSavingsMsg'
Läs mer

2-Chloro-4-(trifluoromethyl)benzenesulfonamide, 97%, Thermo Scientific™

Produktkod. 15438145
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 mg
1 g
Förpackningsstorlek:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15438145 1 g 1g
15448145 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15438145 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H32637.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146533-47-3
Molekylformel C7H5ClF3NO2S
Molekylvikt (g/mol) 259.63
MDL-nummer MFCD00179384
InChI-nyckel ZKBZHFCXIXOBTJ-UHFFFAOYSA-N
Synonym 2-chloro-4-trifluoromethyl benzenesulfonamide, 2-chloro-4-trifluoromethyl benzenesulphonamide, 2-chloro-4-trifluoromethyl benzene sulfonamide, benzenesulfonamide,2-chloro-4-trifluoromethyl, acmc-20dywb, maybridge1_007338, 2-chloro-4-trifluoromethyl-benzenesulfonamide, 2-chloro-4-trifluoromethylbenzene sulfonamide
PubChem CID 2773925
IUPAC-namn 2-chloro-4-(trifluoromethyl)benzenesulfonamide
LEDER NS(=O)(=O)C1=C(Cl)C=C(C=C1)C(F)(F)F

Specifications

Kemiskt namn eller material 2-Chloro-4-(trifluoromethyl)benzenesulfonamide
Smältpunkt 168°C to 171°C
Kvantitet 1 g
Formel vikt 259.63
Procent renhet 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.