Learn More
BRAND™ 2 D Filter Crucible
Filter Crucible constructed of clear glass for visibility.
Brand: BRAND™ 458731
4193.29 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Constructed of Borosilicate Glass 3.3
Specifications
4 cm | |
50 mL | |
1 |
Borosilicate Glass | |
Crucible |
Product Content Correction
Product Title
BRAND™ 2 D Filter Crucible > Porosity: 1
Spot an opportunity for improvement?Share a Content Correction