Läs mer
2-Fluoro-5-methyl-4-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™
Produktkod. 15458965
Change view
Kvantitet:
250 mg
1 g
Förpackningsstorlek:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15458965 250 mg 250mg
15448965 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 15458965 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H34482.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1323966-23-9
Molekylformel C9H7BrF4
Molekylvikt (g/mol) 271.053
MDL-nummer MFCD19687198
InChI-nyckel URFTTWLWYFEASU-UHFFFAOYSA-N
Synonym 2-fluoro-5-methyl-4-trifluoromethyl benzyl bromide, 1-bromomethyl-2-fluoro-5-methyl-4-trifluoromethyl benzene
PubChem CID 66523487
IUPAC-namn 1-(bromomethyl)-2-fluoro-5-methyl-4-(trifluoromethyl)benzene
LEDER CC1=CC(=C(C=C1C(F)(F)F)F)CBr
Specifikationer
Kemiskt namn eller material 2-Fluoro-5-methyl-4-(trifluoromethyl)benzyl bromide
Kvantitet 250 mg
UN-nummer UN3265
Känslighet Moisture sensitive
Formel vikt 271.05
Procent renhet 97%
Fysisk form Liquid