Läs mer
2-Methyl-5-(trifluoromethoxy)phenylacetonitrile, 97%, Thermo Scientific™
Produktkod. 15498445
Change view
Kvantitet:
250 mg
1 g
Förpackningsstorlek:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15498445 250 mg 250mg
15488445 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 15498445 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H33402.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1261683-78-6
Molekylformel C10H8F3NO
Molekylvikt (g/mol) 215.175
MDL-nummer MFCD18399826
InChI-nyckel DSENNMKYJGBGSH-UHFFFAOYSA-N
Synonym 2-methyl-5-trifluoromethoxy phenylacetonitrile, 2-2-methyl-5-trifluoromethoxy phenyl acetonitrile
PubChem CID 86277660
IUPAC-namn 2-[2-methyl-5-(trifluoromethoxy)phenyl]acetonitrile
LEDER CC1=C(C=C(C=C1)OC(F)(F)F)CC#N
Specifikationer
Kemiskt namn eller material 2-Methyl-5-(trifluoromethoxy)phenylacetonitrile
Kvantitet 250 mg
UN-nummer UN3276
Formel vikt 215.17
Procent renhet 97%