Läs mer
2-Methylpyridine-4-boronic acid pinacol ester, 95%, may contain up to 3% water, Thermo Scientific™
Produktkod. 15435125
Change view
Kvantitet:
250 mg
1 g
Förpackningsstorlek:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15435125 250 mg 250mg
11327309 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 15435125 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H27450.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 660867-80-1
Molekylformel C12H18BNO2
Molekylvikt (g/mol) 219.09
MDL-nummer MFCD08061590
InChI-nyckel MBTULFIFECUURA-UHFFFAOYSA-N
Synonym 2-methylpyridine-4-boronic acid pinacol ester, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, 2-methylpyridine-4-boronic acid pinacol, 2-methyl-4-pyridineboronic acid pinacol ester, 2-methylpyridin-4-ylboronic acid pinacol ester, 2-methylpyridin-4-yl boronic acid pinacol ester, 2-methylpyridine-4-yl boronic acid pinacol ester
PubChem CID 16414274
IUPAC-namn 2-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
LEDER CC1=CC(=CC=N1)B1OC(C)(C)C(C)(C)O1
Specifikationer
Kemiskt namn eller material 2-Methylpyridine-4-boronic acid pinacol ester
Smältpunkt 66°C to 71°C
Anteckningar om analysprocent may contain up to 3% water
Kvantitet 250 mg
Formel vikt 219.09
Procent renhet 95%