Läs mer
2-Nonenedioic Acid, TRC
Produktkod. 30008684
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008684 25 mg
30005298 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30008684 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN64954525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 72461-80-4
Molekylformel C9H14O4
Molekylvikt (g/mol) 186.21
Synonym 2-Nonenedioic Acid (Mixture of Z and E Isomers)
IUPAC-namn (E)-non-2-enedioic acid
LEDER O=C(O)CCCCC/C=C/C(O)=O
Specifikationer
Kemiskt namn eller material 2-Nonenedioic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C9H14O4/c10-8(11)6-4-2-1-3-5-7-9(12)13/h4,6H,1-3,5,7H2,(H,10,11)(H,12,13)/b6-4+
Formel vikt 186.0892
Procent renhet >95%