Learn More
Ohaus™ 2 x Centrifuge Rack Adapters, Frontier™ R-S4x250/4MI Swing Out Rotors
Facilitate centrifugation of tubes smaller or greater in number than the original, provided carrier space for Frontier™ R-S4x250/4MI Swing Out Rotors
Brand: Ohaus™ 30314910
3018.71 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Accommodates centrifuge tubes that are smaller or greater in number than the space provided in the rotor carrier
- Suitable for use in Frontier™ R-S4x250/4MI Swing Out Rotors
Specifications
RB | |
24.5 mm | |
Rack, 5x25ml D24.5mm RB, 2/pk | |
2 |
5 x 25 mL | |
R-S4x250/4MI Swing Out Rotors | |
5 |
Product Content Correction
Product Title
Ohaus™ 2 x Centrifuge Rack Adapters, Frontier™ R-S4x250/4MI Swing Out Rotors > 5 x 25 mL
Spot an opportunity for improvement?Share a Content Correction