Learn More
TdB Labs AB 20 kDa Q-dextran for Ion-exchange Chromatography and Biomolecular Studies
Q-dextran is a positively charged dextran modified with quaternary ammonium groups. It is used in ion exchange chromatography, biochemical assays, and for studying electrostatic interactions with negatively charged biomolecules.
619.45 SEK - 5631.01 SEK
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30216325
![]() |
TdB Labs AB
QD20_1g |
1 g |
619.45 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30216524
![]() |
TdB Labs AB
QD20_10g |
10 g |
5631.01 SEK
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Store dry at room temperature, protected from moisture and light.
- Stable for extended periods under recommended storage conditions without loss of charge properties.
Specifications
Modified Dextran |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.