Learn More
Chem Lab 20LT Sulfuric acid 0.25 mol/l (24.52 g H2SO4 / lH2O = 0.5 N (±0.001/20°C)), std sol
Brand: Chem Lab CL05.2614.9520
20LT Sulfuric acid 0.25 mol/l (24.52 g H2SO4 / lH2O = 0.5 N (±0.001/20°C)), std sol
This item is not returnable.
View return policy
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sulphuric acid 0.25 mol/l
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Sulphuric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Chem Lab 20LT Sulfuric acid 0.25 mol/l (24.52 g H2SO4 / lH2O = 0.5 N (±0.001/20°C)), std sol >
Spot an opportunity for improvement?Share a Content Correction