Learn More
Chem Lab 20LT Sulfuric acid 0.25 mol/l (24.52 g H2SO4 / lH2O = 0.5 N (±0.001/20°C)), std sol
Brand:  Chem Lab CL05.2614.9520
20LT Sulfuric acid 0.25 mol/l (24.52 g H2SO4 / lH2O = 0.5 N (±0.001/20°C)), std sol
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11933303
  • 2035.00 SEK / 20 L
Videos
Safety and Handling
Show More
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Chem Lab 20LT Sulfuric acid 0.25 mol/l (24.52 g H2SO4 / lH2O = 0.5 N (±0.001/20°C)), std sol >