Learn More
Thermo Scientific Alfa Aesar 250ML S55603 1323760 - iCAP Q/Qnova CALIBRATION solution
Brand:  Thermo Scientific Alfa Aesar S55603.AK
250ML S55603 1323760 - iCAP Q/Qnova CALIBRATION solution
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery. Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17751488
  • 7860.00 SEK / 250mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1442416 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberS55603.AK
Lot Number1430699 Certificate TypeCertificate of Analysis Date16/05/2024 Catalog NumberS55603.AK
Lot Number1409531 Certificate TypeCertificate of Analysis Date11/04/2024 Catalog NumberS55603.AK
Lot Number1430700 Certificate TypeCertificate of Analysis Date11/04/2024 Catalog NumberS55603.AK
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific Alfa Aesar 250ML S55603 1323760 - iCAP Q/Qnova CALIBRATION solution >