Learn More
3,4 Dinitrotoluene, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep S-1696
Quantity | 1 mL |
---|
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
3,4-Dinitrotoluene | 610-39-9 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
2mL amber prescored glass ampule | |
0.1, 99.9 | |
0.1, 99.9 | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
CC1=CC(=C(C=C1)[N+](=O)[O-])[N+](=O)[O-] | |
Acetonitrile | |
11883 | |
3,4-Dinitrotoluene |
610-39-9, 75-05-8 | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. | |
C7H6N2O4 | |
INYDMNPNDHRJQJ-UHFFFAOYSA-N | |
4-methyl-1,2-dinitrobenzene | |
182.135 | |
1 mL |
Safety and Handling
- 3,4-Dinitrotoluene
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H350-May cause cancer.
- H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : acetonitrile, 3,4-dinitrotoluene