missing translation for 'onlineSavingsMsg'
Learn More

TPP Techno Plastic Products AG 3-B Tissue Culture Flask 150 cm2

3-B Tissue culture flask triple bagged

2355.00 SEK - 8410.00 SEK

Specifications

Capacity (Metric) 15 to 45 mL
Material Polystyrene
Autoclavable Not Autoclavable
Disposable Single-use
Graduated Yes
View More Specs

Products 4
Product Code Brand Closure Type Price Quantity & Availability  
Product Code Brand Closure Type Price Quantity & Availability  
17205306
View Documents
TPP Techno Plastic Products AG
390150
Vented
2355.00 SEK
Pack of 20
Only null left
Estimated Shipment: 03-07-2025
Add to basket
 
17215306
View Documents
TPP Techno Plastic Products AG
390151
Vented Filter cap
2555.00 SEK
Pack of 20
Only null left
Estimated Shipment: 03-07-2025
Add to basket
 
17225306
View Documents
TPP Techno Plastic Products AG
390153
Vented Filter cap, peel-off foil
6875.00 SEK
Pack of 20
Only null left
Estimated Shipment: 17-07-2025
Add to basket
 
17245306
View Documents
TPP Techno Plastic Products AG
390552
Vented Filter cap, re-closeable lid
8410.00 SEK
Pack of 20
Only null left
Estimated Shipment: 03-07-2025
Add to basket
 
Description

Description

  • The triple bagged packaging (3-B) solution from TPP meets the increasingly more stringent clean room requirements.
  • The 3-B products provide noticeably more safety in grade clean aseptic room productions. With the standard packaging, contamination adhering to the outside of the bag could be unintentionally carried into cleanroom areas.
  • The 3-B packaging system prevents contamination from reaching highly sensitive production areas, thus significantly increasing safety for handling substances in a sterile environment.
  • All advantages of the standard product
  • Ideal for use in dedicated areas where the sterile transfer of product is essential
  • Each product is triple-wrapped in three peel-open bags
  • Each bag is marked with a security number: 3 – outer, 2 – middle, 1 – inner bag - the subsequent numbering helps to → show status of packaging
  • Innermost bag 1 is labeled with the product data: product number and description, lot number and EXP.
  • The safety-open-style peel-open bags are increasing sized, so that during handling of one bag the next inner bag is not accidentally broken rendering it contaminated
Specifications

Specifications

15 to 45 mL
Not Autoclavable
Yes
Sterile
150 cm2
Polystyrene
Single-use
Wide
Flat
Triple-bagged
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
TPP Techno Plastic Products AG 3-B Tissue Culture Flask 150 cm2

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.