Läs mer
3-Bromo-4-isopropoxy-5-methoxybenzonitrile, 98%, Thermo Scientific™
Produktkod. 11450120
Change view
Kvantitet:
250 mg
1 g
Förpackningsstorlek:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
11450120 250 mg 250mg
11460120 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 11450120 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H50615.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 515848-62-1
Molekylformel C11H12BrNO2
Molekylvikt (g/mol) 270.13
MDL-nummer MFCD02256549
InChI-nyckel DNCFZWKSHFLLGT-UHFFFAOYSA-N
Synonym 3-bromo-4-isopropoxy-5-methoxybenzonitrile, 3-bromo-5-methoxy-4-propan-2-yloxy benzonitrile, 3-bromo-5-methoxy-4-methylethoxy benzenecarbonitrile, acmc-20an8q, 3-bromo-4-isopropoxy-5-methoxy benzonitrile
PubChem CID 735529
IUPAC-namn 3-bromo-5-methoxy-4-propan-2-yloxybenzonitrile
LEDER COC1=C(OC(C)C)C(Br)=CC(=C1)C#N
Specifikationer
Kemiskt namn eller material 3-Bromo-4-isopropoxy-5-methoxybenzonitrile
Kvantitet 250 mg
UN-nummer UN3439
Formel vikt 270.13
Procent renhet 98%