Läs mer
3-Chloro-4-(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™
Produktkod. 15495057
Change view
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15495057 5 g 5g
15485057 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 15495057 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H32343.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1092460-77-9
Molekylformel C8H3Cl2F3O
Molekylvikt (g/mol) 243.006
MDL-nummer MFCD11519325
InChI-nyckel KKXPAIGCYQCQHM-UHFFFAOYSA-N
Synonym 3-chloro-4-trifluoromethyl benzoyl chloride, 3-chloro-4-trifluoromethyl benzoyl chloride, jrd
PubChem CID 21275239
IUPAC-namn 3-chloro-4-(trifluoromethyl)benzoyl chloride
LEDER C1=CC(=C(C=C1C(=O)Cl)Cl)C(F)(F)F
Specifikationer
Kemiskt namn eller material 3-Chloro-4-(trifluoromethyl)benzoyl chloride
Färg Colorless
Brytningsindex 1.503
Kvantitet 5 g
UN-nummer UN3265
Känslighet Moisture sensitive
Formel vikt 243.01
Procent renhet 97%
Fysisk form Liquid