missing translation for 'onlineSavingsMsg'
Läs mer

3-Chloro-5-fluoro-4-methoxybenzaldehyde, 97%, Thermo Scientific™

Produktkod. 11352579
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11352579 1 g 1g
11362579 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11352579 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H26666.03
Request Bulk or Custom Format

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used as solvents. They are used in intermediate steps for the production of paints, plastics, synthetic resins, and dyes. They are also used in the manufacture of perfumes, solvents, and flavorings.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177994-49-9
Molekylformel C8H6ClFO2
Molekylvikt (g/mol) 188.58
MDL-nummer MFCD04115863
InChI-nyckel HILNMKBXNYUXIJ-UHFFFAOYSA-N
Synonym benzaldehyde,3-chloro-5-fluoro-4-methoxy, acmc-1c8t4, 3-fluoro-5-chloro-4-methoxybenzaldehyde, 5-chloro-3-fluoro-4-methoxybenzaldehyde
PubChem CID 3860780
IUPAC-namn 3-chloro-5-fluoro-4-methoxybenzaldehyde
LEDER COC1=C(F)C=C(C=O)C=C1Cl

Specifications

Kemiskt namn eller material 3-Chloro-5-fluoro-4-methoxybenzaldehyde
Smältpunkt 43°C to 45°C
Kvantitet 1 g
Känslighet Air Sensitive
Löslighetsinformation Insoluble in water.
Formel vikt 188.59
Procent renhet 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.