Läs mer
3-Cysteinylacetaminophen Trifluoroacetic Acid Salt, TRC
Produktkod. 30010861
Change view
Kvantitet:
1 mg
2.5 mg
5 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30010861 2.5 mg 2.5mg
30009016 1 mg -
30011457 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010861 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC9947502.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1331891-93-0
Molekylformel C11 H14 N2 O4 S . C2 H F3 O2
Molekylvikt (g/mol) 384.33
Synonym 3-Cysteinylacetaminophen Tfa Salt
IUPAC-namn (2R)-3-(5-acetamido-2-hydroxyphenyl)sulfanyl-2-aminopropanoic acid;2,2,2-trifluoroacetic acid
LEDER CC(=O)Nc1ccc(O)c(SC[C@H](N)C(=O)O)c1.OC(=O)C(F)(F)F
Specifikationer
Kemiskt namn eller material 3-Cysteinylacetaminophen Trifluoroacetic Acid Salt
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C11H14N2O4S.C2HF3O2/c1-6(14)13-7-2-3-9(15)10(4-7)18-5-8(12)11(16)17;3-2(4,5)1(6)7/h2-4,8,15H,5,12H2,1H3,(H,13,14)(H,16,17);(H,6,7)/t8-;/m0./s1
Kvantitet 2.5 mg
Formel vikt 384.0603
Procent renhet >95%