Läs mer
3-Methoxy-1,2-benzothiazole 1,1-dioxide, TRC
Produktkod. 30468547
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30468547 50 mg
30469168 10 mg
30474248 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30468547 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM26668550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 18712-14-6
Molekylformel C8H7NO3S
Molekylvikt (g/mol) 197.21
Synonym 3-Methoxybenzisothiazole-S,S-dioxide, 3-Methoxybenzo[d]isothiazole 1,1-dioxide, Saccharin O-methyl Ether
IUPAC-namn 3-methoxy-1,2-benzothiazole 1,1-dioxide
LEDER COC1=NS(=O)(=O)c2ccccc12
Specifikationer
Kemiskt namn eller material 3-Methoxy-1,2-benzothiazole 1,1-dioxide
Kvantitet 50 mg
InChI formel InChI=1S/C8H7NO3S/c1-12-8-6-4-2-3-5-7(6)13(10,11)9-8/h2-5H,1H3
Formel vikt 197.0147