Learn More
3-Methyl-3-hexanol, 99% Product Code.: 11371637
3-Methyl-3-hexanol, 99%
Product Code. p-7044105
5 g, 5g
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Product Code. 11371637
Brand: Thermo Scientific Alfa Aesar L04625.06
Request Bulk or Custom Format
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
CAS: 597-96-6 | C7H16O | 116.204 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 597-96-6
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.204
MDL Number MFCD00021839
InChI Key KYWJZCSJMOILIZ-UHFFFAOYSA-N
Synonym 3-methyl-3-hexanol, 3-hexanol, 3-methyl, 2-ethyl-2-pentanol, 3-methyl-hexanol-3, 3-methyl-hexanol-3 german, acmc-20an0r, 4-01-00-01749 beilstein handbook reference
PubChem CID 11708
IUPAC Name 3-methylhexan-3-ol
SMILES CCCC(C)(CC)O
Specifications
Density 0.823
Boiling Point 143°C
Flash Point 45°C (113°F)
Refractive Index 1.4235
Quantity 5 g
UN Number UN1987
Beilstein 1719032
Formula Weight 116.2
Percent Purity 99%
Chemical Name or Material 3-Methyl-3-hexanol
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Product Code
Lot Number10120250 Certificate TypeCertificate of Analysis Date18/03/2024 Product CodeL04625.14,L04625.06
RUO – Research Use Only
Product Content Correction
Product Title
3-Methyl-3-hexanol, 99% >