Läs mer
3-Methyl-4-nitroimino-tetrahydro-1,3,5-oxadiazine, TRC
Produktkod. 30006246
Change view
Kvantitet:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006246 1 g
30004533 250 mg
30006629 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30006246 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3238501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 153719-38-1
Molekylformel C4 H8 N4 O3
Molekylvikt (g/mol) 160.13
Synonym 3-Methyl-4-nitroimino-tetrahydro-1,3,5-oxadiazine
IUPAC-namn N-(3-methyl-1,3,5-oxadiazinan-4-ylidene)nitramide
LEDER CN1COCNC1=N[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material 3-Methyl-4-nitroimino-tetrahydro-1,3,5-oxadiazine
Kvantitet 1 g
InChI formel InChI=1S/C4H8N4O3/c1-7-3-11-2-5-4(7)6-8(9)10/h2-3H2,1H3,(H,5,6)
Formel vikt 160.0596