Läs mer
3-Sulfobenzaldehyde Sodium Salt, TRC
Produktkod. 30132506
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132506 10 g
30004437 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30132506 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS68916510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5330-48-3
Molekylformel C7 H5 O4 S . Na
Molekylvikt (g/mol) 208.17
Synonym m-Benzaldehydesulfonic Acid Sodium Salt, 3-Formylbenzenesulfonic Acid Sodium Salt, Sodium 3-Formylbenzenesulfonate;
IUPAC-namn sodium;3-formylbenzenesulfonate
LEDER [Na+].[O-]S(=O)(=O)c1cccc(C=O)c1
Specifikationer
Kemiskt namn eller material 3-Sulfobenzaldehyde Sodium Salt
Kvantitet 10 g
InChI formel InChI=1S/C7H6O4S.Na/c8-5-6-2-1-3-7(4-6)12(9,10)11;/h1-5H,(H,9,10,11);/q;+1/p-1
Formel vikt 207.9806