Läs mer
(3alpha)-Allopregnanolone 3-beta-D-Glucuronide, TRC
Produktkod. 30136174
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30136174 25 mg -
30008568 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136174 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA54711025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 31300-86-4
Molekylformel C27H42O8
Molekylvikt (g/mol) 494.62
Synonym (3α,5α)-20-Oxopregnan-3-yl β-D-Glucopyranosiduronic Acid, 20-Oxo-5α-pregnan-3α-yl β-D-Glucopyranosiduronic Acid, 20-Oxo-5α-pregnan-3α-yl Glucosiduronic Acid, 3α-Hydroxy-5α-pregnan-20-one Glucuronide;
IUPAC-namn (2S,3S,4S,5R,6R)-6-[[(3R,5S,8R,9S,10S,13S,14S,17S)-17-acetyl-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl]oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid
LEDER CC(=O)[C@H]1CC[C@@H]2[C@@]1(CC[C@H]3[C@H]2CC[C@@H]4[C@@]3(CC[C@H](C4)O[C@H]5[C@@H]([C@H]([C@@H]([C@H](O5)C(=O)O)O)O)O)C)C
Specifikationer
Kemiskt namn eller material (3alpha)-Allopregnanolone 3-beta-D-Glucuronide
Kvantitet 25 mg
InChI formel InChI=1S/C27H42O8/c1-13(28)17-6-7-18-16-5-4-14-12-15(8-10-26(14,2)19(16)9-11-27(17,18)3)34-25-22(31)20(29)21(30)23(35-25)24(32)33/h14-23,25,29-31H,4-12H2,1-3H3,(H,32,33)/t14-,15+,16-,17+,18-,19-,20-,21-,22+,23-,25+,26-,27+/m0/s1
Formel vikt 494.288