Läs mer
4-(2-Methylpropyl)piperidine-2,6-dione, TRC
Produktkod. 17981425
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17981425 250 mg
17960825 100 mg
17912545 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17981425 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM327390250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 916982-10-0
Molekylformel C9 H15 N O2
Molekylvikt (g/mol) 169.22
Synonym 2,6-Piperidinedione, 4-(2-methylpropyl)-, -Isobutylglutarimide, 4-(2-Methylpropyl)piperidine-2,6-dione, 4-Isobutyl-2,6-dioxopiperidine, 4-Isobutylpiperidine-2,6-dione
IUPAC-namn 4-(2-methylpropyl)piperidine-2,6-dione
LEDER CC(C)CC1CC(=O)NC(=O)C1
Specifikationer
Kemiskt namn eller material 4-(2-Methylpropyl)piperidine-2,6-dione
Kvantitet 250 mg
InChI formel InChI=1S/C9H15NO2/c1-6(2)3-7-4-8(11)10-9(12)5-7/h6-7H,3-5H2,1-2H3,(H,10,11,12)
Formel vikt 169.1103