CAS RN 916982-10-0
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
4-(2-Methylpropyl)piperidine-2,6-dione (3-Isobutylglutarimide), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
4-(2-Methylpropyl)piperidine-2,6-dione, TRC
CAS: 916982-10-0 Molekylformel: C9 H15 N O2 Molekylvikt (g/mol): 169.22 Synonym: 2,6-Piperidinedione, 4-(2-methylpropyl)-,-Isobutylglutarimide,4-(2-Methylpropyl)piperidine-2,6-dione,4-Isobutyl-2,6-dioxopiperidine,4-Isobutylpiperidine-2,6-dione IUPAC-namn: 4-(2-methylpropyl)piperidine-2,6-dione LEDER: CC(C)CC1CC(=O)NC(=O)C1