Learn More
Memmert™ 4-20 mA Current Loop Interface for UN75, Actual Value
Temperature Output for external temperature monitoring.
Brand: Memmert™ UN75V3
2496.68 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Temperature controller actual value
Specifications
UN75 Oven | |
Current Loop Interface | |
4 to 20 mA |
Product Content Correction
Product Title
Memmert™ 4-20 mA Current Loop Interface for UN75, Actual Value > For Use With: UN75
Spot an opportunity for improvement?Share a Content Correction