Learn More
Memmert™ 4-20Ma Current Loop Interface Temperature 0-260°C Of One Pt100 In Chamber, For Ich750
Product Code. 16306777 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
ICH750 Climate Chamber
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
16306777 ICH750 Climate Chamber Each
1 options
Product Code. 16306777 Supplier Memmert™ Supplier No. ICH750V6
 Restricted Item
Beräknad leverans: 17-03-2027
Memmert™ Temperature Output for external temperature monitoring.
  • 4 to 20 mA current loop interface
  • Temperature of a Pt100 sensor positioned flexibly in chamber for external temperature monitoring
Specifications
For Use With (Equipment) ICH750 Climate Chamber
Type Current Loop Interface
Amperage 4 to 20 mA