Läs mer
4-Amino Fluconazole Bromide, TRC
Produktkod. 30012750
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012750 25 mg
30011822 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012750 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA63087025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 150168-54-0
Molekylformel C13 H14 F2 N7 O . Br
Molekylvikt (g/mol) 402.2
Synonym 4H-1,2,4-Triazolium, 4-amino-1-[2-(2,4-difluorophenyl)-2-hydroxy-3-(1H-1,2,4-triazol-1-yl)propyl]-, bromide (1:1), 1H-1,2,4-Triazolium, 4-amino-1-[2-(2,4-difluorophenyl)-2-hydroxy-3-(1H-1,2,4-triazol-1-yl)propyl]-, bromide (9CI)
IUPAC-namn 1-(4-amino-1,2,4-triazol-1-ium-1-yl)-2-(2,4-difluorophenyl)-3-(1,2,4-triazol-1-yl)propan-2-ol;bromide
LEDER [Br-].Nn1cn[n+](CC(O)(Cn2cncn2)c3ccc(F)cc3F)c1
Specifikationer
Kemiskt namn eller material 4-Amino Fluconazole Bromide
Kvantitet 25 mg
InChI formel InChI=1S/C13H14F2N7O.BrH/c14-10-1-2-11(12(15)3-10)13(23,4-21-7-17-6-18-21)5-22-9-20(16)8-19-22;/h1-3,6-9,23H,4-5,16H2;1H/q+1;/p-1
Formel vikt 401.0411