Läs mer
4-Chloro-3-iodobenzotrifluoride, 98%, Thermo Scientific™
Produktkod. 11435117
Change view
Kvantitet:
25 g
100 g
Förpackningsstorlek:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
11435117 100 g 100g
11425117 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 11435117 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer B22548.22
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 672-57-1
Molekylformel C7H3ClF3I
Molekylvikt (g/mol) 306.451
MDL-nummer MFCD00001037
InChI-nyckel SSLWFPKNYZEOTH-UHFFFAOYSA-N
Synonym 1-chloro-2-iodo-4-trifluoromethyl benzene, 4-chloro-3-iodobenzotrifluoride, 3-iodo-4-chlorobenzotrifluoride, benzene, 1-chloro-2-iodo-4-trifluoromethyl, 4-chloro-3-iodobenzo trifluoride, pubchem2714, attercop-chm at113459, 1-chloro-2-iodo-4-trifluoromethyl-benzene, 1-chloro-2-iodo-4-trifluoromethyl benzene #
PubChem CID 69596
IUPAC-namn 1-chloro-2-iodo-4-(trifluoromethyl)benzene
LEDER C1=CC(=C(C=C1C(F)(F)F)I)Cl
Specifikationer
Kemiskt namn eller material 4-Chloro-3-iodobenzotrifluoride
Densitet 1.973
Kokpunkt 58°C to 60°C (0.5 mmHg)
Flampunkt >110°C (230°F)
Brytningsindex 1.54
Kvantitet 100 g
Beilstein 1870212
Känslighet Light sensitive
Formel vikt 306.45
Procent renhet 98%
Produkttitel
Välj ett problem
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.