Learn More
4-Cyanobenzoic acid, 99%
CAS: 619-65-8 | C8H5NO2 | 147.13 g/mol
502.00 SEK - 1545.00 SEK
Chemical Identifiers
CAS | 619-65-8 |
---|---|
Molecular Formula | C8H5NO2 |
Molecular Weight (g/mol) | 147.13 |
MDL Number | MFCD00002528 |
InChI Key | ADCUEPOHPCPMCE-UHFFFAOYSA-N |
Synonym | p-cyanobenzoic acid, benzoic acid, 4-cyano, p-carboxybenzonitrile, 4-carboxybenzonitrile, benzoic acid, p-cyano, 4-cyano-benzoic acid, 4-cyano benzoic acid, benzoicacid, 4-cyano, 4-cyanobenzoicacid, para-cyanobenzoic acidShow More |
PubChem CID | 12087 |
IUPAC Name | 4-cyanobenzoic acid |
SMILES | C1=CC(=CC=C1C#N)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10000760
|
Thermo Scientific Acros
110730050 |
5 g | Glass bottle |
502.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10172850
|
Thermo Scientific Acros
110730250 |
25 g | Glass bottle |
1545.00 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
619-65-8 | |
147.13 | |
ADCUEPOHPCPMCE-UHFFFAOYSA-N | |
12087 | |
C1=CC(=CC=C1C#N)C(=O)O |
C8H5NO2 | |
MFCD00002528 | |
p-cyanobenzoic acid, benzoic acid, 4-cyano, p-carboxybenzonitrile, 4-carboxybenzonitrile, benzoic acid, p-cyano, 4-cyano-benzoic acid, 4-cyano benzoic acid, benzoicacid, 4-cyano, 4-cyanobenzoicacid, para-cyanobenzoic acidShow More | |
4-cyanobenzoic acid |
Specifications
619-65-8 | |
100.0 | |
Beige-Yellow to White | |
98.5% min. (HPLC) | |
NCC6H4CO2H | |
09, 845 | |
Solubility in water: 1.2g/L (20°C). Other solubilities: soluble in methanol | |
C1=CC(=CC=C1C#N)C(=O)O | |
147.13 | |
147.13 | |
Crystalline Powder |
98.5 | |
217.0°C to 222.0°C | |
Authentic | |
C8H5NO2 | |
MFCD00002528 | |
p-cyanobenzoic acid, benzoic acid, 4-cyano, p-carboxybenzonitrile, 4-carboxybenzonitrile, benzoic acid, p-cyano, 4-cyano-benzoic acid, 4-cyano benzoic acid, benzoicacid, 4-cyano, 4-cyanobenzoicacid, para-cyanobenzoic acid | |
ADCUEPOHPCPMCE-UHFFFAOYSA-N | |
4-cyanobenzoic acid | |
12087 | |
99% | |
4-Cyanobenzoic acid, 99% |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 210-606-9
missing translation for 'tsca' : TSCA
- 4-Cyanobenzoic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.