missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrophenol, ≥99.5%, Honeywell™ Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
Specifications
Specifications
Melting Point | 116°C |
Boiling Point | 279°C |
Linear Formula | O2NC6H4OH |
UN Number | UN1663 |
Beilstein | 1281877 |
Solubility Information | 14.8g/L at 25°C |
Formula Weight | 139.11g/mol |
Percent Purity | ≥99.5% |
Packaging | Glass bottle |
Vapor Pressure | 0.6mmHg (120°C) |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-Nitrophenol, ≥99.5%, Honeywell™ Fluka™ >
Spot an opportunity for improvement?Share a Content Correction