Läs mer
4-Nitrophenyl 2-Acetamido-6-O-(2-acetamido-2-deoxy-Beta-D-glucopyranosyl) -3-O-(Beta-D-galactopyranosyl)-2-deoxy-Alpha-D-galactopyranoside, TRC
Produktkod. 30474014
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30474014 1 mg
30465499 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30474014 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN4990001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 139459-55-5
Molekylformel C28 H41 N3 O18
Molekylvikt (g/mol) 707.63
IUPAC-namn N-[(2R,3R,4R,5S,6R)-2-[[(2R,3R,4R,5R,6R)-5-acetamido-3-hydroxy-6-(4-nitrophenoxy)-4-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxan-2-yl]methoxy]-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
LEDER CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1OC[C@H]2O[C@H](Oc3ccc(cc3)[N+](=O)[O-])[C@H](NC(=O)C)[C@@H](O[C@@H]4O[C@H](CO)[C@H](O)[C@H](O)[C@H]4O)[C@H]2O
Specifikationer
Kemiskt namn eller material 4-Nitrophenyl 2-Acetamido-6-O-(2-acetamido-2-deoxy-beta-D-glucopyranosyl) -3-O-(beta-D-galactopyranosyl)-2-deoxy-alpha-D-galactopyranoside
Kvantitet 1 mg
InChI formel InChI=1S/C28H41N3O18/c1-10(34)29-17-22(39)19(36)14(7-32)46-26(17)44-9-16-21(38)25(49-28-24(41)23(40)20(37)15(8-33)47-28)18(30-11(2)35)27(48-16)45-13-5-3-12(4-6-13)31(42)43/h3-6,14-28,32-33,36-41H,7-9H2,1-2H3,(H,29,34)(H,30,35)/t14-,15-,16-,17-,18-,19-,20+,21+,22-,23+,24-,25-,26-,27+,28+/m1/s1
Formel vikt 707.2385