Läs mer
4-(Pentafluorothio)benzyl alcohol, 97%, Thermo Scientific™
Produktkod. 15437337
Change view
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15437337 1 g 1g
15447337 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 15437337 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H34397.03
Request Bulk or Custom Format
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 773872-73-4
Molekylformel C7H7F5OS
Molekylvikt (g/mol) 234.184
MDL-nummer MFCD06203063
InChI-nyckel QKEQGUCFBIRBAU-UHFFFAOYSA-N
Synonym 4-pentafluorosulfur benzyl alcohol, 4-pentafluoro-??-sulfanyl phenyl methanol, 4-pentafluorothio benzyl alcohol
PubChem CID 58360645
IUPAC-namn [4-(pentafluoro-$l^{6}-sulfanyl)phenyl]methanol
LEDER C1=CC(=CC=C1CO)S(F)(F)(F)(F)F
Specifikationer
Kemiskt namn eller material 4-(Pentafluorothio)benzyl alcohol
Kvantitet 1 g
Formel vikt 234.18
Procent renhet 97%