Läs mer
(4R)-1-Methyl-4-propyl-L-proline Hydrochloride, TRC
Produktkod. 30005335
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005335 100 mg
30010057 50 mg
17979254 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30005335 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM320285100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6734-79-8
Molekylformel C9 H17 N O2 . Cl H
Molekylvikt (g/mol) 207.7
Synonym L-trans-1-Methyl-4-propyl-proline Hydrochloride, 4-n-Propyl Hygric Acid Hydrochloride, trans-Propylhygric Acid Hydrochloride, 1-Methyl-4-propyl-L-trans-proline Hydrochloride
IUPAC-namn (2S,4R)-1-methyl-4-propylpyrrolidine-2-carboxylic acid;hydrochloride
LEDER Cl.CCC[C@@H]1C[C@H](N(C)C1)C(=O)O
Specifikationer
Kemiskt namn eller material (4R)-1-Methyl-4-propyl-L-proline Hydrochloride
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C9H17NO2.ClH/c1-3-4-7-5-8(9(11)12)10(2)6-7;/h7-8H,3-6H2,1-2H3,(H,11,12);1H/t7-,8+;/m1./s1
Kvantitet 100 mg
Formel vikt 207.1026
Procent renhet >95%