missing translation for 'onlineSavingsMsg'
Läs mer

(4R)-1-Methyl-4-propyl-L-proline Hydrochloride, TRC

Produktkod. 30010057
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010057 50 mg
30005335 100 mg
17979254 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30010057 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32028550MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 6734-79-8
Molekylformel C9 H17 N O2 . Cl H
Molekylvikt (g/mol) 207.7
Synonym L-trans-1-Methyl-4-propyl-proline Hydrochloride, 4-n-Propyl Hygric Acid Hydrochloride, trans-Propylhygric Acid Hydrochloride, 1-Methyl-4-propyl-L-trans-proline Hydrochloride
IUPAC-namn (2S,4R)-1-methyl-4-propylpyrrolidine-2-carboxylic acid;hydrochloride
LEDER Cl.CCC[C@@H]1C[C@H](N(C)C1)C(=O)O

Specifikationer

Kemiskt namn eller material (4R)-1-Methyl-4-propyl-L-proline Hydrochloride
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C9H17NO2.ClH/c1-3-4-7-5-8(9(11)12)10(2)6-7;/h7-8H,3-6H2,1-2H3,(H,11,12);1H/t7-,8+;/m1./s1
Kvantitet 50 mg
Formel vikt 207.1026
Procent renhet >95%
Frakt skick Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.