Läs mer
5-Desmethyl-3-methyl Leflunomide, TRC
Produktkod. 30358100
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30358100 100 mg
30351975 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30358100 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD291935100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 208401-20-1
Molekylformel C12 H9 F3 N2 O2
Molekylvikt (g/mol) 270.21
Synonym 3-Methyl-N-[4-(trifluoromethyl)phenyl]isoxazole-4-carboxamide, 4-Isoxazolecarboxamide, 3-methyl-N-[4-(trifluoromethyl)phenyl]- (9CI), 5-Desmethyl-3-methyl Leflunomide
IUPAC-namn 3-methyl-N-[4-(trifluoromethyl)phenyl]-1,2-oxazole-4-carboxamide
LEDER Cc1nocc1C(=O)Nc2ccc(cc2)C(F)(F)F
Specifikationer
Kemiskt namn eller material 3-Methyl-N-[4-(trifluoromethyl)phenyl]isoxazole-4-carboxamide
Kvantitet 100 mg
InChI formel InChI=1S/C12H9F3N2O2/c1-7-10(6-19-17-7)11(18)16-9-4-2-8(3-5-9)12(13,14)15/h2-6H,1H3,(H,16,18)
Formel vikt 270.0616