CAS RN 208401-20-1
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
2
av
2
Resultat
1
3-Methyl-N-[4-(trifluoromethyl)phenyl]isoxazole-4-carboxamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
5-Desmethyl-3-methyl Leflunomide, TRC
CAS: 208401-20-1 Molekylformel: C12 H9 F3 N2 O2 Molekylvikt (g/mol): 270.21 Synonym: 3-Methyl-N-[4-(trifluoromethyl)phenyl]isoxazole-4-carboxamide,4-Isoxazolecarboxamide, 3-methyl-N-[4-(trifluoromethyl)phenyl]- (9CI),5-Desmethyl-3-methyl Leflunomide IUPAC-namn: 3-methyl-N-[4-(trifluoromethyl)phenyl]-1,2-oxazole-4-carboxamide LEDER: Cc1nocc1C(=O)Nc2ccc(cc2)C(F)(F)F