Läs mer
5-Hydroxymethyl-2-furoylglycine, TRC
Produktkod. 17990335
Change view
Kvantitet:
5 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17990335 5 mg
17912155 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17990335 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9727805MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 38716-68-6
Molekylformel C8 H9 N O5
Molekylvikt (g/mol) 199.161
Synonym N-[[5-(Hydroxymethyl)-2-furanyl]carbonyl]glycine, 5-(hydroxymethyl)-promucuric Acid
IUPAC-namn 2-[[5-(hydroxymethyl)furan-2-carbonyl]amino]acetic acid
LEDER OCc1oc(cc1)C(=O)NCC(=O)O
Specifikationer
Kemiskt namn eller material 5-Hydroxymethyl-2-furoylglycine
InChI formel InChI=1S/C8H9NO5/c10-4-5-1-2-6(14-5)8(13)9-3-7(11)12/h1-2,10H,3-4H2,(H,9,13)(H,11,12)
Kvantitet 5 mg
Formel vikt 199.0481