Learn More
Reagecon 500ML pH Buffer Solution pH 1.07 ± 0.05 @25°C-Clear(for use with Antimony or equivalent electrodes)
BIOPHARMA
Brand:  Reagecon 10725050
500ML pH Buffer Solution pH 1.07 ± 0.05 @25°C-Clear(for use with Antimony or equivalent electrodes)
Product Code. 16512370
  • 461.00 SEK / 500mL
Videos
Safety and Handling
Show More
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1725023E1 Certificate TypeCertificate of Analysis Date15/03/2024 Catalog Number10725050
Promotions
Promotions
Product Content Correction
Product Title
Reagecon 500ML pH Buffer Solution pH 1.07 ± 0.05 @25°C-Clear(for use with Antimony or equivalent electrodes) >