Learn More
DiscoverX [D-Ala2, D-Leu5]-Enkephalin (DADLE)
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
[D-Ala2, D-Leu5]-Enkephalin (DADLE) is a synthetic peptide that is a selective agonist of the delta opioid receptor, but also binds mu opioid receptors of subtype ÎĽ1. It has antinociceptive and analgesic properties via neuron protection, and in-vivo effects include reduction in arterial blood pressure and heart rate. Provided as 5mg of dry powder alongside reconstitution buffer.
Specifications
Specifications
Product Type | Control Ligands |
Quantity | 5 mg |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
DiscoverX [D-Ala2, D-Leu5]-Enkephalin (DADLE)
Spot an opportunity for improvement?Share a Content Correction